missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpentane 98.0+%, TCI America™

Catalog No. p-7132132
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M038225ML 25 mL
M0382500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M038225ML Supplier TCI America Supplier No. M038225ML
Only null left

Chemical Identifiers

CAS 107-83-5
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00009406
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID 7892
IUPAC Name 2-methylpentane
SMILES CCCC(C)C

Specifications

Color Colorless
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 1208
Boiling Point 60°C
Formula Weight 86.18
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Methylpentane
EINECSNumber (2)-0006
RTECSNumber SA2985000
TSCA Yes