Learn More
2-Methylpentane, 99+%
CAS: 107-83-5 | C6H14 | 86.178 g/mol
Supplier: Thermo Scientific Chemicals L03001AP
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications2-Methylpentane is employed as a raw material, rubber solvent and vegetable oil extraction solvent. It is also used as a solvent in adhesives. Further, it is used as an intermediate in organic synthesis and finds application in food, preservatives, cosmetics, pharmaceuticals, beverages and flavor enhancer.
Solubility
Miscible with alcohol, ether, acetone and chloroform. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| 2-Methylpentane | |
| -154°C | |
| 61°C to 62°C | |
| Petroleum-like | |
| 1.371 | |
| 500 mL | |
| 1730735 | |
| Miscible with alcohol,ether,acetone and chloroform. Immiscible with water. | |
| CCCC(C)C | |
| 86.178 | |
| 86.18 |
| 107-83-5 | |
| 0.653 | |
| −40°C (−40°F) | |
| C6H14 | |
| MFCD00009406 | |
| UN1208 | |
| isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
| AFABGHUZZDYHJO-UHFFFAOYSA-N | |
| 2-methylpentane | |
| 7892 | |
| ≥99% |
Chemical Identifiers
| 107-83-5 | |
| 86.178 | |
| AFABGHUZZDYHJO-UHFFFAOYSA-N | |
| 7892 | |
| CCCC(C)C |
| C6H14 | |
| MFCD00009406 | |
| isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
| 2-methylpentane |
Safety and Handling
GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES
EINECSNumber : 203-523-4
RTECSNumber : SA2985000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only