missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpentane-2,4-diol 99.0+%, TCI America™

Catalog No. p-7132134
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M038425ML 25 mL
M0384500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M038425ML Supplier TCI America Supplier No. M038425ML
Only null left

Chemical Identifiers

CAS 107-41-5
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00004547
InChI Key SVTBMSDMJJWYQN-UHFFFAOYSA-N
Synonym hexylene glycol, 2-methyl-2,4-pentanediol, pinakon, 2,4-dihydroxy-2-methylpentane, 2,4-pentanediol, 2-methyl, diolane, isol, 4-methyl-2,4-pentanediol, 1,1,3-trimethyltrimethylenediol, 2-methyl pentane-2,4-diol
PubChem CID 7870
ChEBI CHEBI:62995
IUPAC Name 2-methylpentane-2,4-diol
SMILES CC(CC(C)(C)O)O

Specifications

Color Colorless
Boiling Point 197°C
Quantity 25 mL
Formula Weight 118.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylpentane-2,4-diol
EINECSNumber (2)-0240
RTECSNumber SA0810000
TSCA Yes