missing translation for 'onlineSavingsMsg'
Learn More

2-Methylnonane 99.0+%, TCI America™

Catalog No. m02811g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M02811G Supplier TCI America Supplier No. M02811G
Only null left

Chemical Identifiers

CAS 871-83-0
Molecular Formula C10H22
Molecular Weight (g/mol) 142.29
MDL Number MFCD00027321
InChI Key SGVYKUFIHHTIFL-UHFFFAOYSA-N
Synonym nonane, 2-methyl, c9-11 isoparaffin, alkanes, c9-11-iso, 2-methylnonane, unii-382akh052v, isodecane, octane, dimethyl, iso-decane, 2-methyl nonane, c10-13 isoparaffin
PubChem CID 13379
IUPAC Name 2-methylnonane
SMILES CCCCCCCC(C)C

Specifications

Color Colorless
Boiling Point 166°C
Quantity 1 g
UN Number 3295
Formula Weight 142.29
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylnonane
EINECSNumber (2)-0010
TSCA No