missing translation for 'onlineSavingsMsg'
Learn More

2-Methylnonane, 98+%

CAS: 871-83-0 | C10H22 | 142.29 g/mol

Supplier:  Thermo Scientific Chemicals B2084906

 View more versions of this product

Catalog No. AAB2084906

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methylnonane
0.72
46°C (115°F)
C10H22
CH3(CH2)6CH(CH3)2
5 g
1696990
SGVYKUFIHHTIFL-UHFFFAOYSA-N
2-methylnonane
13379
≥98%
871-83-0
166°C
Petroleum-like
1.41
MFCD00027321
UN3295
nonane, 2-methyl, c9-11 isoparaffin, alkanes, c9-11-iso, 2-methylnonane, unii-382akh052v, isodecane, octane, dimethyl, iso-decane, 2-methyl nonane, c10-13 isoparaffin
CCCCCCCC(C)C
142.29
142.29

Chemical Identifiers

871-83-0
142.29
SGVYKUFIHHTIFL-UHFFFAOYSA-N
13379
CCCCCCCC(C)C
C10H22
MFCD00027321
nonane, 2-methyl, c9-11 isoparaffin, alkanes, c9-11-iso, 2-methylnonane, unii-382akh052v, isodecane, octane, dimethyl, iso-decane, 2-methyl nonane, c10-13 isoparaffin
2-methylnonane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c

H226-H304

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 212-814-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only