missing translation for 'onlineSavingsMsg'
Learn More

2-Methylnonane, 98+%

Catalog No. AAB2084914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2084914 25 g
AAB2084903 1 g
AAB2084906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2084914 Supplier Thermo Scientific Chemicals Supplier No. B2084914

CAS: 871-83-0 | C10H22 | 142.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 871-83-0
Molecular Formula C10H22
Molecular Weight (g/mol) 142.29
MDL Number MFCD00027321
InChI Key SGVYKUFIHHTIFL-UHFFFAOYSA-N
Synonym nonane, 2-methyl, c9-11 isoparaffin, alkanes, c9-11-iso, 2-methylnonane, unii-382akh052v, isodecane, octane, dimethyl, iso-decane, 2-methyl nonane, c10-13 isoparaffin
PubChem CID 13379
IUPAC Name 2-methylnonane
SMILES CCCCCCCC(C)C

Specifications

Density 0.72
Boiling Point 166°C
Flash Point 46°C (115°F)
Odor Petroleum-like
Refractive Index 1.41
Linear Formula CH3(CH2)6CH(CH3)2
Quantity 25 g
UN Number UN3295
Beilstein 1696990
Formula Weight 142.29
Percent Purity ≥98%
Chemical Name or Material 2-Methylnonane
Show More Show Less
Hazard Category H226-H304
Hazard Statement GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 212-814-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only