missing translation for 'onlineSavingsMsg'
Learn More

2-Methylnicotinic Acid 98.0+%, TCI America™

Catalog No. M20891G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20891G 1 g
M20895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20891G Supplier TCI America Supplier No. M20891G

Chemical Identifiers

CAS 3222-56-8
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00013449
InChI Key HNTZKNJGAFJMHQ-UHFFFAOYSA-N
Synonym 2-methylnicotinic acid, 2-methyl-nicotinic acid, 2-methylnicotinicacid, 3-pyridinecarboxylic acid, 2-methyl, 2-methyl nicotinic acid, 2-methyl-3-pyridinecarboxylic acid, methylnicotinic acid, pubchem2458, 2-methylnicotinoic acid, acmc-1ctxp
PubChem CID 643373
IUPAC Name 2-methylpyridine-3-carboxylic acid
SMILES CC1=C(C=CC=N1)C(=O)O

Specifications

Melting Point 227°C
Color White-Yellow
Quantity 1 g
Formula Weight 137.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylnicotinic Acid