Learn More
2-Methylnicotinic acid, 98+%, Thermo Scientific™
Chemical Identifiers
| CAS | 3222-56-8 |
|---|---|
| Molecular Formula | C7H7NO2 |
| Molecular Weight (g/mol) | 137.14 |
| MDL Number | MFCD00013449 |
| InChI Key | HNTZKNJGAFJMHQ-UHFFFAOYSA-N |
| Synonym | 2-methylnicotinic acid, 2-methyl-nicotinic acid, 2-methylnicotinicacid, 3-pyridinecarboxylic acid, 2-methyl, 2-methyl nicotinic acid, 2-methyl-3-pyridinecarboxylic acid, methylnicotinic acid, pubchem2458, 2-methylnicotinoic acid, acmc-1ctxp |
| PubChem CID | 643373 |
| IUPAC Name | 2-methylpyridine-3-carboxylic acid |
| SMILES | CC1=C(C=CC=N1)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC397320010
|
Thermo Scientific Chemicals
397320010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC397320050
|
Thermo Scientific Chemicals
397320050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC397320250
|
Thermo Scientific Chemicals
397320250 |
25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 3222-56-8 | |
| 137.14 | |
| HNTZKNJGAFJMHQ-UHFFFAOYSA-N | |
| 643373 | |
| CC1=C(C=CC=N1)C(=O)O |
| C7H7NO2 | |
| MFCD00013449 | |
| 2-methylnicotinic acid, 2-methyl-nicotinic acid, 2-methylnicotinicacid, 3-pyridinecarboxylic acid, 2-methyl, 2-methyl nicotinic acid, 2-methyl-3-pyridinecarboxylic acid, methylnicotinic acid, pubchem2458, 2-methylnicotinoic acid, acmc-1ctxp | |
| 2-methylpyridine-3-carboxylic acid |
Specifications
| 3222-56-8 | |
| 100.0 | |
| 98% min. (HPLC) | |
| C7H7NO2 | |
| 1g | |
| HNTZKNJGAFJMHQ-UHFFFAOYSA-N | |
| 2-methylpyridine-3-carboxylic acid | |
| 643373 | |
| 98+% |
| 98.0 | |
| Authentic | |
| Glass bottle | |
| MFCD00013449 | |
| 2-methylnicotinic acid, 2-methyl-nicotinic acid, 2-methylnicotinicacid, 3-pyridinecarboxylic acid, 2-methyl, 2-methyl nicotinic acid, 2-methyl-3-pyridinecarboxylic acid, methylnicotinic acid, pubchem2458, 2-methylnicotinoic acid, acmc-1ctxp | |
| CC1=C(C=CC=N1)C(=O)O | |
| 137.14 | |
| 137.14 | |
| 2-Methylnicotinic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO â Research Use Only