missing translation for 'onlineSavingsMsg'
Learn More

2-Methylnicotinic acid, 98+%

Catalog No. AC397320250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC397320250 25 g
AC397320010 1 g
AC397320050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC397320250 Supplier Thermo Scientific Chemicals Supplier No. 397320250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3222-56-8 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3222-56-8
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00013449
InChI Key HNTZKNJGAFJMHQ-UHFFFAOYSA-N
Synonym 2-methylnicotinic acid, 2-methyl-nicotinic acid, 2-methylnicotinicacid, 3-pyridinecarboxylic acid, 2-methyl, 2-methyl nicotinic acid, 2-methyl-3-pyridinecarboxylic acid, methylnicotinic acid, pubchem2458, 2-methylnicotinoic acid, acmc-1ctxp
PubChem CID 643373
IUPAC Name 2-methylpyridine-3-carboxylic acid
SMILES CC1=C(C=CC=N1)C(=O)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 225.0°C to 228.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Quantity 25 g
Formula Weight 137.14
Percent Purity 98+%
Physical Form Crystals or Powder
Chemical Name or Material 2-Methylnicotinic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only