missing translation for 'onlineSavingsMsg'
Learn More

2-Methylnaphth[2,1-d]oxazole 98.0+%, TCI America™

Catalog No. p-7133293
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25591G 1 g
M25595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25591G Supplier TCI America Supplier No. M25591G
Only null left

Chemical Identifiers

CAS 20686-65-1
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00022185
InChI Key JKYSRHYQAVELLH-UHFFFAOYSA-N
Synonym 2-methylnaphth 2,1-d oxazole, 2-methylnaphtho 2,1-d oxazole, naphth 2,1-d oxazole, 2-methyl, 2-methylnaphtho 2,1-d 1,3 oxazole, 2-methylbenzo g 1,3 benzoxazole, pubchem8703, 2-methylbenzo g benzoxazole
PubChem CID 88652
IUPAC Name 2-methylbenzo[g][1,3]benzoxazole
SMILES CC1=NC2=C(O1)C3=CC=CC=C3C=C2

Specifications

Melting Point 41°C
Color White-Yellow
Boiling Point 302°C
Quantity 1 g
Formula Weight 183.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylnaphth[2,1-d]oxazole