missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylindole-3-carboxaldehyde 98.0+%, TCI America™
$95.06 - $294.52
Chemical Identifiers
| CAS | 5416-80-8 |
|---|---|
| Molecular Formula | C10H9NO |
| Molecular Weight (g/mol) | 159.188 |
| MDL Number | MFCD00012077 |
| InChI Key | CYZIVXOEJNAIBS-UHFFFAOYSA-N |
| Synonym | 2-methylindole-3-carboxaldehyde, 3-formyl-2-methylindole, 2-methyl-3-formylindole, 1h-indole-3-carboxaldehyde, 2-methyl, 2-methylindole-3-carbaldehyde, indole-3-carboxaldehyde, 2-methyl, pubchem7437, zlchem 1339, acmc-1apkw, ksc271e8j |
| PubChem CID | 73166 |
| IUPAC Name | 2-methyl-1H-indole-3-carbaldehyde |
| SMILES | CC1=C(C2=CC=CC=C2N1)C=O |
Chemical Identifiers
| 5416-80-8 | |
| 159.188 | |
| CYZIVXOEJNAIBS-UHFFFAOYSA-N | |
| 73166 | |
| CC1=C(C2=CC=CC=C2N1)C=O |
| C10H9NO | |
| MFCD00012077 | |
| 2-methylindole-3-carboxaldehyde, 3-formyl-2-methylindole, 2-methyl-3-formylindole, 1h-indole-3-carboxaldehyde, 2-methyl, 2-methylindole-3-carbaldehyde, indole-3-carboxaldehyde, 2-methyl, pubchem7437, zlchem 1339, acmc-1apkw, ksc271e8j | |
| 2-methyl-1H-indole-3-carbaldehyde |
Specifications
| 5416-80-8 | |
| C10H9NO | |
| 5 g | |
| CYZIVXOEJNAIBS-UHFFFAOYSA-N | |
| 2-methyl-1H-indole-3-carbaldehyde | |
| 73166 | |
| ≥98.0% (GC) | |
| 2-Methylindole-3-carboxaldehyde |
| 203°C | |
| MFCD00012077 | |
| 2-methylindole-3-carboxaldehyde, 3-formyl-2-methylindole, 2-methyl-3-formylindole, 1h-indole-3-carboxaldehyde, 2-methyl, 2-methylindole-3-carbaldehyde, indole-3-carboxaldehyde, 2-methyl, pubchem7437, zlchem 1339, acmc-1apkw, ksc271e8j | |
| CC1=C(C2=CC=CC=C2N1)C=O | |
| 159.188 | |
| 159.19 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0136
TSCA : Yes