missing translation for 'onlineSavingsMsg'
Learn More

2-Methylindole-3-carboxaldehyde 98.0+%, TCI America™

Catalog No. M14855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14855G 5 g
M148525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14855G Supplier TCI America Supplier No. M14855G

Chemical Identifiers

CAS 5416-80-8
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00012077
InChI Key CYZIVXOEJNAIBS-UHFFFAOYSA-N
Synonym 2-methylindole-3-carboxaldehyde, 3-formyl-2-methylindole, 2-methyl-3-formylindole, 1h-indole-3-carboxaldehyde, 2-methyl, 2-methylindole-3-carbaldehyde, indole-3-carboxaldehyde, 2-methyl, pubchem7437, zlchem 1339, acmc-1apkw, ksc271e8j
PubChem CID 73166
IUPAC Name 2-methyl-1H-indole-3-carbaldehyde
SMILES CC1=C(C2=CC=CC=C2N1)C=O

Specifications

Melting Point 203°C
Quantity 5 g
Formula Weight 159.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylindole-3-carboxaldehyde
EINECSNumber (5)-0136
TSCA Yes