missing translation for 'onlineSavingsMsg'
Learn More

2-Methylindole-3-carboxaldehyde, 97+%, Thermo Scientific™

Chemical Identifiers

CAS 5416-80-8
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00012077
InChI Key CYZIVXOEJNAIBS-UHFFFAOYSA-N
Synonym 2-methylindole-3-carboxaldehyde, 3-formyl-2-methylindole, 2-methyl-3-formylindole, 1h-indole-3-carboxaldehyde, 2-methyl, 2-methylindole-3-carbaldehyde, indole-3-carboxaldehyde, 2-methyl, pubchem7437, zlchem 1339, acmc-1apkw, ksc271e8j
PubChem CID 73166
IUPAC Name 2-methyl-1H-indole-3-carbaldehyde
SMILES CC1=C(C2=CC=CC=C2N1)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC379720250
SDS View Documents
Thermo Scientific Chemicals
379720250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC379721000
SDS View Documents
Thermo Scientific Chemicals
379721000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5416-80-8
159.19
CYZIVXOEJNAIBS-UHFFFAOYSA-N
73166
CC1=C(C2=CC=CC=C2N1)C=O
C10H9NO
MFCD00012077
2-methylindole-3-carboxaldehyde, 3-formyl-2-methylindole, 2-methyl-3-formylindole, 1h-indole-3-carboxaldehyde, 2-methyl, 2-methylindole-3-carbaldehyde, indole-3-carboxaldehyde, 2-methyl, pubchem7437, zlchem 1339, acmc-1apkw, ksc271e8j
2-methyl-1H-indole-3-carbaldehyde

Specifications

5416-80-8
100.0
97% min. (HPLC)
C10H9NO
25g
CYZIVXOEJNAIBS-UHFFFAOYSA-N
2-methyl-1H-indole-3-carbaldehyde
73166
97+%
97.0
Authentic
Glass bottle
MFCD00012077
2-methylindole-3-carboxaldehyde, 3-formyl-2-methylindole, 2-methyl-3-formylindole, 1h-indole-3-carboxaldehyde, 2-methyl, 2-methylindole-3-carbaldehyde, indole-3-carboxaldehyde, 2-methyl, pubchem7437, zlchem 1339, acmc-1apkw, ksc271e8j
CC1=C(C2=CC=CC=C2N1)C=O
159.19
159.19
2-Methylindole-3-carboxaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-512-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only