missing translation for 'onlineSavingsMsg'
Learn More

2-Methylindole-3-acetic acid, 98+%

Catalog No. AAL1907103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1907103 1 g
AAL1907106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1907103 Supplier Thermo Scientific Chemicals Supplier No. L1907103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1912-43-2 | C11H11NO2 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1912-43-2
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00075006
InChI Key QJNNHJVSQUUHHE-UHFFFAOYSA-N
Synonym 2-methylindole-3-acetic acid, 2-methyl-1h-indol-3-yl acetic acid, 2-2-methyl-1h-indol-3-yl acetic acid, 2-methylindole-3-aceticacid, 1h-indole-3-acetic acid, 2-methyl, 2-methyl-3-indoleacetic acid, 2-2-methylindol-3-yl acetic acid, pubchem7229, acmc-209euw, 2-methylindole-3acetic acid
PubChem CID 589107
IUPAC Name 2-(2-methyl-1H-indol-3-yl)acetic acid
SMILES CC1=C(CC(O)=O)C2=CC=CC=C2N1

Specifications

Melting Point 198°C to 204°C (decomposition)
Quantity 1 g
Beilstein 152701
Formula Weight 189.22
Percent Purity ≥98%
Chemical Name or Material 2-Methylindole-3-acetic acid
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only