missing translation for 'onlineSavingsMsg'
Learn More

2-Methylindene, Thermo Scientific Chemicals

Catalog No. AC467760010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467760010 1 g
AC467760050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467760010 Supplier Thermo Scientific Chemicals Supplier No. 467760010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2177-47-1
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD00274253
InChI Key YSAXEHWHSLANOM-UHFFFAOYSA-N
IUPAC Name 2-methyl-1H-indene
SMILES CC1=CC2=CC=CC=C2C1

Specifications

Density 0.9710 g/mL
Flash Point 76°C
Quantity 1 g
Specific Gravity 0.971
Formula Weight 130.19
Chemical Name or Material 2-Methylindene

RUO – Research Use Only