missing translation for 'onlineSavingsMsg'
Learn More

2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC21701DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC21701DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC21701DA Supplier Thermo Scientific Chemicals Supplier No. CC21701DA
Only null left

Chemical Identifiers

CAS 21801-79-6
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.18
MDL Number MFCD00244615
InChI Key FBIUGCLQMKPURJ-UHFFFAOYSA-N
Synonym 2-methylimidazo 1,2-a pyridine-3-carboxylic acid, 2-methyl-imidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 2-methyl, 2-methyl-4-hydroimidazo 1,2-a pyridine-3-carboxylic acid, pubchem16305, imidazo 1, 2-methyl, 2-methylimidazo 1,2-a nicotinic acid, 3-carboxy-2-methylimidazo 1,2-a pyridine
PubChem CID 89054
IUPAC Name 2-methylimidazo[1,2-a]pyridine-3-carboxylic acid
SMILES CC1=C(N2C=CC=CC2=N1)C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material 2-Methylimidazo[1, 2-a]pyridine-3-carboxylic acid
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.