missing translation for 'onlineSavingsMsg'
Learn More

2-Methylhexanoyl chloride, 97%

Catalog No. AAH3282909
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3282909 10 g
AAH3282903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3282909 Supplier Thermo Scientific Chemicals Supplier No. H3282909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 41693-47-4 | C7H13ClO | 148.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41693-47-4
Molecular Formula C7H13ClO
Molecular Weight (g/mol) 148.63
MDL Number MFCD01318644
InChI Key WKEBLSOTVYFYKY-UHFFFAOYNA-N
Synonym 2-methylcaproyl chloride, 2-methyl hexanoyl chloride, hexanoyl chloride,2-methyl
PubChem CID 3458543
IUPAC Name 2-methylhexanoyl chloride
SMILES CCCCC(C)C(Cl)=O

Specifications

Density 0.957
Boiling Point 163°C
Flash Point 55°C (131°F)
Refractive Index 1.429
Quantity 10 g
UN Number UN2920
Sensitivity Moisture sensitive
Formula Weight 148.63
Percent Purity 97%
Chemical Name or Material 2-Methylhexanoyl chloride
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only