missing translation for 'onlineSavingsMsg'
Learn More

2-Methylheptanoic Acid 98.0+%, TCI America™

Supplier:  TCI America M126525G

 View more versions of this product

Catalog No. M126525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Methylheptanoic Acid
Yellow
C8H16O2
25 g
2-methylenanthic acid, heptanoic acid, 2-methyl, methylamylacetic acid, 2-methyl-heptanoic acid, heptanoic acid, methyl, alpha-methylheptanoic acid, 2-methyl heptanoic acid, heptane-2-carboxylic acid, fema no. 2706, heptanoic acid, 2-methyl-, 2s
CCCCCC(C)C(=O)O
144.214
144.21
Liquid
1188-02-9
140°C
MFCD00142944
3265
NKBWMBRPILTCRD-UHFFFAOYSA-N
2-methylheptanoic acid
14475
≥98.0% (T,GC)

Chemical Identifiers

1188-02-9
144.214
NKBWMBRPILTCRD-UHFFFAOYSA-N
14475
CCCCCC(C)C(=O)O
C8H16O2
MFCD00142944
2-methylenanthic acid, heptanoic acid, 2-methyl, methylamylacetic acid, 2-methyl-heptanoic acid, heptanoic acid, methyl, alpha-methylheptanoic acid, 2-methyl heptanoic acid, heptane-2-carboxylic acid, fema no. 2706, heptanoic acid, 2-methyl-, 2s
2-methylheptanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0608

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions