missing translation for 'onlineSavingsMsg'
Learn More

2-Methylheptanoic Acid 98.0+%, TCI America™

Catalog No. M12655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12655G 5 g
M126525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12655G Supplier TCI America Supplier No. M12655G

Chemical Identifiers

CAS 1188-02-9
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00142944
InChI Key NKBWMBRPILTCRD-UHFFFAOYSA-N
Synonym 2-methylenanthic acid, heptanoic acid, 2-methyl, methylamylacetic acid, 2-methyl-heptanoic acid, heptanoic acid, methyl, alpha-methylheptanoic acid, 2-methyl heptanoic acid, heptane-2-carboxylic acid, fema no. 2706, heptanoic acid, 2-methyl-, 2s
PubChem CID 14475
IUPAC Name 2-methylheptanoic acid
SMILES CCCCCC(C)C(=O)O

Specifications

Color Yellow
Boiling Point 140°C
Quantity 5 g
UN Number 3265
Formula Weight 144.21
Percent Purity ≥98.0% (T,GC)
Physical Form Liquid
Chemical Name or Material 2-Methylheptanoic Acid
EINECSNumber (2)-0608
TSCA Yes