missing translation for 'onlineSavingsMsg'
Learn More

2-Methylheptanoic acid, 98%

Catalog No. AAB2205006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2205006 5 g
AAB2205014 25 g
AAB2205022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2205006 Supplier Thermo Scientific Chemicals Supplier No. B2205006
Only null left

CAS: 1188-02-9 | C8H16O2 | 144.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1188-02-9
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00142944
InChI Key NKBWMBRPILTCRD-UHFFFAOYSA-N
Synonym 2-methylenanthic acid, heptanoic acid, 2-methyl, methylamylacetic acid, 2-methyl-heptanoic acid, heptanoic acid, methyl, alpha-methylheptanoic acid, 2-methyl heptanoic acid, heptane-2-carboxylic acid, fema no. 2706, heptanoic acid, 2-methyl-, 2s
PubChem CID 14475
IUPAC Name 2-methylheptanoic acid
SMILES CCCCCC(C)C(=O)O

Specifications

Density 0.906
Boiling Point 139°C to 141°C (30 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4255
Quantity 5 g
Formula Weight 144.22
Percent Purity 98%
Chemical Name or Material 2-Methylheptanoic acid
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 214-704-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only