missing translation for 'onlineSavingsMsg'
Learn More

2-Methylheptane, 99%

CAS: 592-27-8 | C8H18 | 114.23 g/mol

$206.82 - $850.86

Chemical Identifiers

CAS 592-27-8
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number MFCD00009518
InChI Key JVSWJIKNEAIKJW-UHFFFAOYSA-N
Synonym heptane, 2-methyl, methylheptane, 2-methyl-heptane, unii-yu6su8ccvb, yu6su8ccvb, hexane, dimethyl, dimethylhexane, acmc-1avkm, 2-methylheptane, ch3 2ch ch2 4ch3
PubChem CID 11594
IUPAC Name 2-methylheptane
SMILES CCCCCC(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC126880050
SDS View Documents
Thermo Scientific Chemicals
126880050
5 g Glass bottle
Each for $206.82
Only null left
 
AC126880250
SDS View Documents
Thermo Scientific Chemicals
126880250
25 g Glass bottle
Each for $850.86
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

592-27-8
114.23
JVSWJIKNEAIKJW-UHFFFAOYSA-N
11594
CCCCCC(C)C
C8H18
MFCD00009518
heptane, 2-methyl, methylheptane, 2-methyl-heptane, unii-yu6su8ccvb, yu6su8ccvb, hexane, dimethyl, dimethylhexane, acmc-1avkm, 2-methylheptane, ch3 2ch ch2 4ch3
2-methylheptane

Specifications

592-27-8
100.0
Colorless
116°C (761.0 mmHg)
Authentic
Glass bottle
1.3940 to 1.396
MFCD00009518
01, 161
heptane, 2-methyl, methylheptane, 2-methyl-heptane, unii-yu6su8ccvb, yu6su8ccvb, hexane, dimethyl, dimethylhexane, acmc-1avkm, 2-methylheptane, ch3 2ch ch2 4ch3
JVSWJIKNEAIKJW-UHFFFAOYSA-N
2-methylheptane
11594
99%
2-Methylheptane, 99%
98.5
-108.9°C
0.6900g/mL
4°C
98.5% min. (GC)
C8H18
CH3(CH2)4CH(CH3)2
5 g
0.69
Solubility in water: insoluble. Other solubilities: soluble in ether,miscible with ethanol,acetone
CCCCCC(C)C
114.23
114.23
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-747-9

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only