missing translation for 'onlineSavingsMsg'
Learn More

2-Methylglutaronitrile 98.0+%, TCI America™

Catalog No. p-7132102
Change view
Click to view available options
Quantity:
10 g
25 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M033110G 10 g
M033125G 25 g
M0331500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M033110G Supplier TCI America Supplier No. M033110G
Only null left

Chemical Identifiers

CAS 4553-62-2
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00001875
InChI Key FPPLREPCQJZDAQ-UHFFFAOYSA-N
Synonym 1,3-Dicyanobutane
PubChem CID 20686
IUPAC Name 2-methylpentanedinitrile
SMILES CC(CCC#N)C#N

Specifications

Color Yellow
Boiling Point 135°C
Quantity 10 g
UN Number 3276
Formula Weight 108.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylglutaronitrile
RTECSNumber YV8140000
TSCA Yes