missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylglutaronitrile 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 135°C |
| Quantity | 25 g |
| UN Number | 3276 |
| Formula Weight | 108.14 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Methylglutaronitrile |
Safety and Handling
| RTECSNumber | YV8140000 |
| TSCA | Yes |