missing translation for 'onlineSavingsMsg'
Learn More

2'-Methylformanilide 98.0+%, TCI America™

$44.28 - $144.39

Chemical Identifiers

CAS 94-69-9
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00014122
InChI Key ZXTLGJAARBNQGK-UHFFFAOYSA-N
Synonym 2'-methylformanilide, 2-methylformanilide, o-tolylformamide, n-2-methylphenyl formamide, formamide, n-2-methylphenyl, o-methylformanilide, o-formotoluidide, o-methyl-n-formylaniline, 2-methylphenylformamide, n-formyl-2-methylaniline
PubChem CID 7202
IUPAC Name N-(2-methylphenyl)formamide
SMILES CC1=CC=CC=C1NC=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M14315G
View Documents
TCI America
M14315G
5 g
Each for $44.28
Only null left
 
M143125G
View Documents
TCI America
M143125G
25 g
Each for $144.39
Only null left
 
Description

Description

Specifications

Chemical Identifiers

94-69-9
135.17
ZXTLGJAARBNQGK-UHFFFAOYSA-N
7202
CC1=CC=CC=C1NC=O
C8H9NO
MFCD00014122
2'-methylformanilide, 2-methylformanilide, o-tolylformamide, n-2-methylphenyl formamide, formamide, n-2-methylphenyl, o-methylformanilide, o-formotoluidide, o-methyl-n-formylaniline, 2-methylphenylformamide, n-formyl-2-methylaniline
N-(2-methylphenyl)formamide

Specifications

94-69-9
C8H9NO
5 g
ZXTLGJAARBNQGK-UHFFFAOYSA-N
N-(2-methylphenyl)formamide
7202
≥98.0% (GC)
2′-Methylformanilide
61°C
MFCD00014122
2'-methylformanilide, 2-methylformanilide, o-tolylformamide, n-2-methylphenyl formamide, formamide, n-2-methylphenyl, o-methylformanilide, o-formotoluidide, o-methyl-n-formylaniline, 2-methylphenylformamide, n-formyl-2-methylaniline
CC1=CC=CC=C1NC=O
135.17
135.17
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents