missing translation for 'onlineSavingsMsg'
Learn More

2'-Methylformanilide 98.0+%, TCI America™

Catalog No. M14315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14315G 5 g
M143125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14315G Supplier TCI America Supplier No. M14315G

Chemical Identifiers

CAS 94-69-9
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00014122
InChI Key ZXTLGJAARBNQGK-UHFFFAOYSA-N
Synonym 2'-methylformanilide, 2-methylformanilide, o-tolylformamide, n-2-methylphenyl formamide, formamide, n-2-methylphenyl, o-methylformanilide, o-formotoluidide, o-methyl-n-formylaniline, 2-methylphenylformamide, n-formyl-2-methylaniline
PubChem CID 7202
IUPAC Name N-(2-methylphenyl)formamide
SMILES CC1=CC=CC=C1NC=O

Specifications

Melting Point 61°C
Quantity 5 g
Formula Weight 135.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Methylformanilide