missing translation for 'onlineSavingsMsg'
Learn More

2-Methylene-1,3-propanediol, 97%

Catalog No. AC308590010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC308590010 1 g Glass bottle
AC308590050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC308590010 Supplier Thermo Scientific Chemicals Supplier No. 308590010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3513-81-3 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3513-81-3
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00075162
InChI Key JFFYKITVXPZLQS-UHFFFAOYSA-N
Synonym 2-methylene-1,3-propanediol, 2-methylenepropane-1,3-diol, 1,3-propanediol, 2-methylene, 1,3-dihydroxy-2-methylenepropane, 1,3-dihydroxy-2-methylidenepropane, dimethylol ethylene, 2-methylene1,3-propanediol, 2-methylenepropan-1,3-diol, ksc573o4t, 1,3-propanediol,2-methylene
PubChem CID 77048
IUPAC Name 2-methylidenepropane-1,3-diol
SMILES C=C(CO)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 93°C to 95°C (2.0 mmHg)
Flash Point <110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4730 to 1.475
Linear Formula HOCH2C(=CH2)CH2OH
Quantity 1 g
Specific Gravity 1.08
Formula Weight 88.11
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material 2-Methylene-1, 3-propanediol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 222-516-7

RUO – Research Use Only