missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclopropanecarboxylic acid, cis + trans, 96%

Catalog No. AAH2781509
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2781509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2781509 Supplier Thermo Scientific Chemicals Supplier No. H2781509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 29555-02-0 | C5H7O2 | 99.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29555-02-0
Molecular Formula C5H7O2
Molecular Weight (g/mol) 99.11
MDL Number MFCD00001293
InChI Key AYEGPMGNMOIHDL-IUYQGCFVSA-M
Synonym 2-methylcyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 2-methyl, 2-methyl-1-cyclopropanecarboxylic acid, trans-2-methylcyclopropanecarboxylic acid, cis-2-methylcyclopropanecarboxylic acid, acmc-1cg8e, 2-methyl-cyclopropanecarboxylic acid, 2-methyl cyclopropane carboxylic acid, cyclopropanecarboxylicacid, 2-methyl
PubChem CID 99873
IUPAC Name 2-methylcyclopropane-1-carboxylic acid
SMILES C[C@H]1C[C@H]1C([O-])=O

Specifications

Density 1.027
Boiling Point 200°C to 203°C
Flash Point 88°C (190°F)
Refractive Index 1.439
Quantity 10 g
UN Number UN3265
Beilstein 2323873
Formula Weight 100.12
Percent Purity 97%
Chemical Name or Material 2-Methylcyclopropanecarboxylic acid, Cis + Trans
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 249-693-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only