missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylcyclopenta[l]phenanthrene 97.0+%, TCI America™
$219.95 - $1109.11
Chemical Identifiers
| CAS | 121254-39-5 |
|---|---|
| Molecular Formula | C18H14 |
| Molecular Weight (g/mol) | 230.31 |
| MDL Number | MFCD03844801 |
| InChI Key | YODBCXSJSTXEIB-UHFFFAOYSA-N |
| PubChem CID | 15842243 |
| IUPAC Name | 2-methyl-1H-cyclopenta[l]phenanthrene |
| SMILES | CC1=CC2=C(C1)C3=CC=CC=C3C4=CC=CC=C42 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M1411100MG
|
TCI America
M1411100MG |
100 mg |
Each for $219.95
|
|
|||||
|
M14111G
|
TCI America
M14111G |
1 g |
Each for $1,109.11
|
|
|||||
Chemical Identifiers
| 121254-39-5 | |
| 230.31 | |
| YODBCXSJSTXEIB-UHFFFAOYSA-N | |
| 2-methyl-1H-cyclopenta[l]phenanthrene |
| C18H14 | |
| MFCD03844801 | |
| 15842243 | |
| CC1=CC2=C(C1)C3=CC=CC=C3C4=CC=CC=C42 |
Specifications
| 121254-39-5 | |
| C18H14 | |
| 100 mg | |
| CC1=CC2=C(C1)C3=CC=CC=C3C4=CC=CC=C42 | |
| 230.31 | |
| 230.31 | |
| Crystalline Powder |
| 166°C | |
| MFCD03844801 | |
| YODBCXSJSTXEIB-UHFFFAOYSA-N | |
| 2-methyl-1H-cyclopenta[l]phenanthrene | |
| 15842243 | |
| ≥97.0% (GC) | |
| 2-Methylcyclopenta[l]phenanthrene |
Safety and Handling
TSCA : No