missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexylamine (cis- and trans- mixture) 95.0+%, TCI America™

Catalog No. M0896500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0896500ML 500 mL
M089625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0896500ML Supplier TCI America Supplier No. M0896500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7003-32-9
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.20
MDL Number MFCD00001492
InChI Key FEUISMYEFPANSS-UHFFFAOYNA-N
Synonym 2-methylcyclohexylamine, cyclohexanamine, 2-methyl, o-methylcyclohexylamine, cyclohexylamine, 2-methyl, 2-methylcyclohexanamine, 2-methylcyclohexylamine,c&t, hexahydro-o-toluidine, 2methylcyclohexylamine, 2-methyl-cyclohexylamine, 2-methylcyclohexyl amine
PubChem CID 23432
IUPAC Name 2-methylcyclohexan-1-amine
SMILES CC1CCCCC1N

Specifications

Color Yellow
Quantity 500 mL
UN Number 2733
Formula Weight 113.20
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methylcyclohexylamine (cis- and trans- mixture)
EINECSNumber (3)-3249&(9)-2279
TSCA Yes