missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexylamine, cis + trans, 97%

Catalog No. p-7043715
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0228509 10 g
AAL0228518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0228509 Supplier Thermo Scientific Chemicals Supplier No. L0228509

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7003-32-9 | C7H15N | 113.20 g/mol

2-Methylcyclohexylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.

Chemical Identifiers

CAS 7003-32-9
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.20
MDL Number MFCD00001492
InChI Key FEUISMYEFPANSS-UHFFFAOYNA-N
Synonym 2-methylcyclohexylamine, cyclohexanamine, 2-methyl, o-methylcyclohexylamine, cyclohexylamine, 2-methyl, 2-methylcyclohexanamine, 2-methylcyclohexylamine,c&t, hexahydro-o-toluidine, 2methylcyclohexylamine, 2-methyl-cyclohexylamine, 2-methylcyclohexyl amine
PubChem CID 23432
IUPAC Name 2-methylcyclohexan-1-amine
SMILES CC1CCCCC1N

Specifications

Density 0.858
Boiling Point 149°C to 150°C
Flash Point 21°C (69°F)
Refractive Index 1.4565
Quantity 10 g
UN Number UN2733
Beilstein 2689073
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 113.2
Percent Purity 97%
Chemical Name or Material 2-Methylcyclohexylamine, Cis + Trans
Show More Show Less
Hazard Category H225-H302+H312+H332-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 230-277-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only