missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexyl Acetate 97.0+%, TCI America™

Catalog No. A132925ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A132925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A132925ML Supplier TCI America Supplier No. A132925ML

Chemical Identifiers

CAS 5726-19-2
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.23
MDL Number MFCD00021390
InChI Key AKIIJALHGMKJEJ-UHFFFAOYNA-N
Synonym Acetic Acid 2-Methylcyclohexyl Ester
PubChem CID 98345
IUPAC Name 2-methylcyclohexyl acetate
SMILES CC1CCCCC1OC(C)=O

Specifications

Color Yellow
Boiling Point 182°C
Quantity 25 mL
Formula Weight 156.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylcyclohexyl Acetate
EINECSNumber (3)-2311&(3)-2356
TSCA No