missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexanone, 98%

Catalog No. p-7022120
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA14741AE 100 mL
AAA14741AP 500 mL
AAA147410F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA14741AE Supplier Thermo Scientific Chemicals Supplier No. A14741AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 583-60-8 | C7H12O | 112.172 g/mol

2-Methylcyclohexanone is used as a fragrant and flavoring agent. It can also be used for the preparation of a novel pladienolide analog scaffold. Also used as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexanone is used as a fragrant and flavoring agent. It can also be used for the preparation of a novel pladienolide analog scaffold. Also used as a solvent.

Solubility
Insoluble in water. Soluble in methanol and diethyl ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 583-60-8
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00001635
InChI Key LFSAPCRASZRSKS-UHFFFAOYSA-N
Synonym 2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone
PubChem CID 11419
IUPAC Name 2-methylcyclohexan-1-one
SMILES CC1CCCCC1=O

Specifications

Melting Point °C to 14°C
Density 0.925
Boiling Point 162°C to 163°C
Flash Point 46°C (114°F)
Assay Percent Range 98%
Refractive Index 1.448
Quantity 100 mL
UN Number UN2297
Beilstein 506751
Solubility Information Insoluble in water. Soluble in methanol and diethyl ether.
Formula Weight 112.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methylcyclohexanone
Show More Show Less
Hazard Category H226-H332
Hazard Statement GHS H Statement
H226-H332
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHYLCYCLOHEXANONE
EINECSNumber 209-513-6
RTECSNumber GW1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only