missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexanone 96.0+%, TCI America™

Catalog No. p-7132048
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M019725ML 25 mL
M0197100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M019725ML Supplier TCI America Supplier No. M019725ML

Chemical Identifiers

CAS 583-60-8
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00001635
InChI Key LFSAPCRASZRSKS-UHFFFAOYSA-N
Synonym 2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone
PubChem CID 11419
IUPAC Name 2-methylcyclohexan-1-one
SMILES CC1CCCCC1=O

Specifications

Color Colorless
Boiling Point 165°C
Quantity 25 mL
UN Number 2297
Formula Weight 112.17
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylcyclohexanone
EINECSNumber (3)-2411
RTECSNumber GW1750000
TSCA Yes