missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylcyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™
$41.02 - $165.90
Chemical Identifiers
| CAS | 583-59-5 |
|---|---|
| Molecular Formula | C7H14O |
| Molecular Weight (g/mol) | 114.188 |
| MDL Number | MFCD00001443 |
| InChI Key | NDVWOBYBJYUSMF-UHFFFAOYSA-N |
| Synonym | 2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers |
| PubChem CID | 11418 |
| IUPAC Name | 2-methylcyclohexan-1-ol |
| SMILES | CC1CCCCC1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M019425ML
|
TCI America
M019425ML |
25 mL |
Each for $41.02
|
|
|||||
|
M0194500ML
|
TCI America
M0194500ML |
500 mL |
Each for $165.90
|
|
|||||
Chemical Identifiers
| 583-59-5 | |
| 114.188 | |
| NDVWOBYBJYUSMF-UHFFFAOYSA-N | |
| 11418 | |
| CC1CCCCC1O |
| C7H14O | |
| MFCD00001443 | |
| 2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers | |
| 2-methylcyclohexan-1-ol |
Specifications
| 583-59-5 | |
| C7H14O | |
| 25 mL | |
| 2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers | |
| CC1CCCCC1O | |
| 114.188 | |
| 114.19 | |
| Liquid |
| Colorless | |
| MFCD00001443 | |
| 2617 | |
| NDVWOBYBJYUSMF-UHFFFAOYSA-N | |
| 2-methylcyclohexan-1-ol | |
| 11418 | |
| ≥98.0% (GC) | |
| 2-Methylcyclohexanol (cis- and trans- mixture) |
Safety and Handling
EINECSNumber : (3)-2301
RTECSNumber : GW0220000
TSCA : Yes