missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. M0194500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0194500ML 500 mL
M019425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0194500ML Supplier TCI America Supplier No. M0194500ML

Chemical Identifiers

CAS 583-59-5
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00001443
InChI Key NDVWOBYBJYUSMF-UHFFFAOYSA-N
Synonym 2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers
PubChem CID 11418
IUPAC Name 2-methylcyclohexan-1-ol
SMILES CC1CCCCC1O

Specifications

Color Colorless
Quantity 500 mL
UN Number 2617
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylcyclohexanol (cis- and trans- mixture)
EINECSNumber (3)-2301
RTECSNumber GW0220000
TSCA Yes