Learn More
2-Methylcyclohexanol, cis + trans, 97%
CAS: 583-59-5 | C7H14O | 114.188 g/mol
Supplier: Thermo Scientific Chemicals A128770F
Description
2-Methylcyclohexanol is used in the preparation of acetic acid-(2-methyl-cyclohexyl ester) by reaction with acetic anhydride. Further, it is used to study the effect of organic solvents on epoxide hydrolase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methylcyclohexanol is used in the preparation of acetic acid-(2-methyl-cyclohexyl ester) by reaction with acetic anhydride. Further, it is used to study the effect of organic solvents on epoxide hydrolase.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| 2-Methylcyclohexanol | |
| 583-59-5 | |
| 0.929 | |
| 58°C (136°F) | |
| 1.462 | |
| 2500 mL | |
| 1847535 | |
| Slightly miscible with water. | |
| CC1CCCCC1O | |
| 114.188 | |
| 114.19 |
| cis + trans | |
| -21°C | |
| 163°C to 166°C | |
| C7H14O | |
| MFCD00001443 | |
| UN2617 | |
| 2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers | |
| NDVWOBYBJYUSMF-UHFFFAOYSA-N | |
| 2-methylcyclohexan-1-ol | |
| 11418 | |
| 97% |
Chemical Identifiers
| 583-59-5 | |
| 114.188 | |
| NDVWOBYBJYUSMF-UHFFFAOYSA-N | |
| 11418 | |
| CC1CCCCC1O |
| C7H14O | |
| MFCD00001443 | |
| 2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers | |
| 2-methylcyclohexan-1-ol |
Safety and Handling
GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
H226-H302+H332-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHYLCYCLOHEXANOLS
EINECSNumber : 209-512-0
RTECSNumber : GW0220000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only