missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcinnamic Acid 98.0+%, TCI America™

Catalog No. p-7132648
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12955G 5 g
M129525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12955G Supplier TCI America Supplier No. M12955G

Chemical Identifiers

CAS 2373-76-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00016841
InChI Key RSWBWHPZXKLUEX-VOTSOKGWSA-N
Synonym 2-methylcinnamic acid, 3-o-tolyl acrylic acid, o-methylcinnamic acid, 2-methylcinnamic acid, predominantly trans, 2e-3-2-methylphenyl acrylic acid, e-3-2-methylphenyl-2-propenoic acid, 2e-3-2-methylphenyl prop-2-enoic acid, e-3-o-tolyl acrylic acid, e-3-o-tolylacrylic acid, 3-o-tolyl-acrylic acid
PubChem CID 819020
IUPAC Name (E)-3-(2-methylphenyl)prop-2-enoic acid
SMILES CC1=CC=CC=C1C=CC(=O)O

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 5 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylcinnamic Acid