missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylbutyric acid, 98%
Description
2-Methylbutyric acid, can be used as food additive for butter, cream, cheese flavor deployment. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used in the synthesis of 2-methylbutyric anhydride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methylbutyric acid, can be used as food additive for butter, cream, cheese flavor deployment. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used in the synthesis of 2-methylbutyric anhydride.
Solubility
It is soluble in water, but highly soluble in most common organic solvents such as Chloroform, Methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Bases, Oxidizing agents, Reducing agents.
2-Methylbutyric acid, can be used as food additive for butter, cream, cheese flavor deployment. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used in the synthesis of 2-methylbutyric anhydride.
Solubility
It is soluble in water, but highly soluble in most common organic solvents such as Chloroform, Methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Bases, Oxidizing agents, Reducing agents.
Specifications
Specifications
| Density | 0.937 |
| Boiling Point | 175°C to 176°C |
| Flash Point | 73°C (163°F) |
| Assay Percent Range | 98% |
| Refractive Index | 1.4055 |
| Quantity | 25 mL |
| UN Number | UN3265 |
| Beilstein | 1720486 |
| Solubility Information | It is soluble in water,but highly soluble in most common organic solvents such as Chloroform,Methanol. |
| Formula Weight | 102.14 |
| Show More |
Safety and Handling
| Hazard Category | H227-H302+H312-H314 |
| Hazard Statement | GHS H Statement H301-H311-H314-H318-H227 Toxic if swallowed. Toxic in contact with skin. Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
| Precautionary Statement | P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 204-145-2 |
| RTECSNumber | EK7897000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only