missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutyraldehyde 95.0+%, TCI America™

Catalog No. M016625ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M016625ML 25 mL
M0166100ML 100 mL
M0166500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M016625ML Supplier TCI America Supplier No. M016625ML

Chemical Identifiers

CAS 96-17-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00006984
InChI Key BYGQBDHUGHBGMD-UHFFFAOYNA-N
Synonym 2-methylbutyraldehyde, butanal, 2-methyl, 2-formylbutane, 2-methylbutyric aldehyde, methylethylacetaldehyde, 2-ethylpropanal, 2-methyl-1-butanal, butyraldehyde, 2-methyl, acetaldehyde, ethylmethyl, alpha-methylbutanal
PubChem CID 7284
ChEBI CHEBI:16182
IUPAC Name 2-methylbutanal
SMILES CCC(C)C=O

Specifications

Color Colorless
Boiling Point 94°C
Quantity 25 mL
UN Number 3371
Formula Weight 86.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylbutyraldehyde
EINECSNumber (2)-0494
RTECSNumber ES3400000
TSCA Yes
Recommended Storage Refrigerator