missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutyraldehyde, 95%

Catalog No. AC304472500
Change view
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC304472500 250 mL Glass bottle
AC304470500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC304472500 Supplier Thermo Scientific Chemicals Supplier No. 304472500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 96-17-3 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96-17-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00006984
InChI Key BYGQBDHUGHBGMD-UHFFFAOYNA-N
Synonym 2-methylbutyraldehyde, butanal, 2-methyl, 2-formylbutane, 2-methylbutyric aldehyde, methylethylacetaldehyde, 2-ethylpropanal, 2-methyl-1-butanal, butyraldehyde, 2-methyl, acetaldehyde, ethylmethyl, alpha-methylbutanal
PubChem CID 7284
ChEBI CHEBI:16182
IUPAC Name 2-methylbutanal
SMILES CCC(C)C=O

Specifications

Acidity 10mg KOH/g max.
CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 0.8050g/mL
Boiling Point 90°C to 92°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH(CH3)CHO
Refractive Index 1.3860 to 1.394
Quantity 250 mL
Beilstein 01, I, 352
Specific Gravity 0.805
Solubility Information Solubility in water: 17g/L (20°C). Other solubilities: soluble in alcohol and ether
Viscosity 0.6 mPa.s (20°C)
Formula Weight 86.13
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Methylbutyraldehyde, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation or rash occurs: Get medical a
EINECSNumber 202-485-6
RTECSNumber ES3400000
TSCA TSCA

RUO – Research Use Only