missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylbutylamine 98.0+%, TCI America™
$180.81 - $501.43
Chemical Identifiers
| CAS | 96-15-1 |
|---|---|
| Molecular Formula | C5H13N |
| Molecular Weight (g/mol) | 87.166 |
| MDL Number | MFCD00008147 |
| InChI Key | VJROPLWGFCORRM-UHFFFAOYSA-N |
| Synonym | 2-methylbutylamine, 1-butanamine, 2-methyl, 1-amino-2-methylbutane, 2-methylbutyl amine, 2-methyl-1-butylamine, 2-methylbutanamine, 2-methyl-1-butanamine, butylamine, 2-methyl, 1-amino-2-methyl-butane, 2-methyl-butylamine |
| PubChem CID | 7283 |
| IUPAC Name | 2-methylbutan-1-amine |
| SMILES | CCC(C)CN |
Chemical Identifiers
| 96-15-1 | |
| 87.166 | |
| VJROPLWGFCORRM-UHFFFAOYSA-N | |
| 7283 | |
| CCC(C)CN |
| C5H13N | |
| MFCD00008147 | |
| 2-methylbutylamine, 1-butanamine, 2-methyl, 1-amino-2-methylbutane, 2-methylbutyl amine, 2-methyl-1-butylamine, 2-methylbutanamine, 2-methyl-1-butanamine, butylamine, 2-methyl, 1-amino-2-methyl-butane, 2-methyl-butylamine | |
| 2-methylbutan-1-amine |
Specifications
| 96-15-1 | |
| 97°C | |
| MFCD00008147 | |
| 2733 | |
| VJROPLWGFCORRM-UHFFFAOYSA-N | |
| 2-methylbutan-1-amine | |
| 7283 | |
| ≥98.0% (GC) | |
| 2-Methylbutylamine |
| Yellow | |
| C5H13N | |
| 5 mL | |
| 2-methylbutylamine, 1-butanamine, 2-methyl, 1-amino-2-methylbutane, 2-methylbutyl amine, 2-methyl-1-butylamine, 2-methylbutanamine, 2-methyl-1-butanamine, butylamine, 2-methyl, 1-amino-2-methyl-butane, 2-methyl-butylamine | |
| CCC(C)CN | |
| 87.166 | |
| 87.17 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0133
TSCA : Yes