missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutyl Acetate 98.0+%, TCI America™

Catalog No. p-7117229
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A107625ML 25 mL
A1076500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A107625ML Supplier TCI America Supplier No. A107625ML
Only null left

Chemical Identifiers

CAS 624-41-9
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00040494
InChI Key XHIUFYZDQBSEMF-UHFFFAOYSA-N
Synonym Acetic Acid 2-Methylbutyl Ester
PubChem CID 12209
ChEBI CHEBI:50585
IUPAC Name 2-methylbutyl acetate
SMILES CCC(C)COC(=O)C

Specifications

Color Colorless
Boiling Point 141°C
Quantity 25 mL
UN Number 1104
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylbutyl Acetate
EINECSNumber (2)-0733
RTECSNumber EL5466666
TSCA Yes