missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutyl Acetate 98.0+%, TCI America™

Catalog No. a1076500ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
A1076500ML 500 mL
A107625ML 25 mL
2 options

Catalog No. A1076500ML

Supplier: TCI America A1076500ML

Only null left

Chemical Identifiers

CAS 624-41-9
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00040494
InChI Key XHIUFYZDQBSEMF-UHFFFAOYSA-N
Synonym Acetic Acid 2-Methylbutyl Ester
PubChem CID 12209
ChEBI CHEBI:50585
IUPAC Name 2-methylbutyl acetate
SMILES CCC(C)COC(=O)C

Specifications

Color Colorless
Boiling Point 141°C
Quantity 500 mL
UN Number 1104
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylbutyl Acetate
EINECSNumber (2)-0733
RTECSNumber EL5466666
TSCA Yes