missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutane 97.0+%, TCI America™

Catalog No. M016725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M016725ML 25 mL
M0167500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M016725ML Supplier TCI America Supplier No. M016725ML

Chemical Identifiers

CAS 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009338
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
PubChem CID 6556
ChEBI CHEBI:30362
IUPAC Name 2-methylbutane
SMILES CCC(C)C

Specifications

Color Colorless
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 1265
Boiling Point 31°C
Formula Weight 72.15
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Methylbutane
EINECSNumber (2)-0005
RTECSNumber EK4430000
TSCA Yes