missing translation for 'onlineSavingsMsg'
Learn More

StatLab 2-Methylbutane (Isopentane) Reagent

Catalog No. 21402154
Change view
Click to view available options
Color:
Colorless
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21402154 Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21402154 Supplier StatLab Supplier No. SPM1543
Only null left
Harmful if swallowed, in contact with skin, or if inhaled

A mastertech special stain kit component used for cryosectioning

  • Solution is a significant component of natural gasoline and is used in conjunction with dry ice or liquid nitrogen to achieve a liquid bath temperature of 160° C
  • Used in conjunction with dry ice or liquid nitrogen to freeze tissues for cryosectioning in histology
  • A MasterTech Special Stain Kit Component

Specifications

Physical Form Solution
Color Colorless
CAS 78-78-4
Quantity 500 mL
Grade Laboratory