missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutane, MilliporeSigma™

Catalog No. M1060561000
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1060561000 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1060561000 Supplier Technidata Supplier No. 1.06056.1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation

Chemical Identifiers

CAS 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009338
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
PubChem CID 6556
ChEBI CHEBI:30362
IUPAC Name 2-methylbutane
SMILES CCC(C)C

Specifications

Color Colorless
Density 0.62g/cm3 (20°C)
Boiling Point 28°C
Flash Point -57°C (70.6°F)
Odor Characteristic
Packaging Glass Bottle
Quantity 1 L
UN Number UN 1265
Solubility Information Water: 0.048g/L at 48°C (118°F)
Vapor Pressure 800 hPa (20°C)
Formula Weight 72.15
Physical Form Liquid
Chemical Name or Material 2-Methylbutane
Show More Show Less
Hazard Category H224: Extremely flammable liquid and vapor.
Hazard Statement H304: May be fatal if swallowed and enters airways.
Precautionary Statement H336: May cause drowsiness or dizziness.
EINECSNumber 201-142-8
RTECSNumber EK4430000
Recommended Storage <15°C (59°F); Keep away from heat and sources of ignition. Keep container tightly closed in a dry and well ventilated place.