missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutane, CHROMASOLV™, for HPLC, ≥99.5%, Honeywell Riedel-de Haën™

Catalog No. p-7111781
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000877 1 L Glass Bottle
6000878 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000877 Supplier Honeywell Chemicals Supplier No. 2703421L
Only null left

CHROMASOLV™, for HPLC,≥99.5%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Chemical Identifiers

CAS 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009338
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
PubChem CID 6556
ChEBI CHEBI:30362
IUPAC Name 2-methylbutane
SMILES CCC(C)C

Specifications

Absorbance max. 0.01 at 400 nm, max. 0.01 at 240 nm, max. 0.07 at 220 nm, max. 0.30 at 210 nm, max. 1.0 at 192 nm
CAS Max % 100.0000%
Density 0.62 g/mL at 25 °C(lit.)
Boiling Point 28°C
Flash Point −51°C
Packaging Glass Bottle
Linear Formula CH3CH2CH(CH3)2
Quantity 1 L
UN Number UN1265
Beilstein 1730723
Formula Weight 72.15
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material 2-Methylbutane, CHROMASOLV™, for HPLC, ≥99.5%
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Pentanes
EINECSNumber 201-142-8
Recommended Storage Room Temp