missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylbutane, 99+%, extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Melting Point | -160.0°C |
| Color | Colorless |
| Density | 0.6200g/mL |
| Boiling Point | 30.0°C |
| Flash Point | −51°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99+% |
| Packaging | Glass Bottle |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Danger |
| missing translation for 'healthHazard2' | GHS H Statement: May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. May be fatal if swallowed and enters airways. Extremely flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking. |
| missing translation for 'healthHazard3' | GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Take precautionary measures against static discharge. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only