missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutane, 98%, Spectrum™ Chemical

Catalog No. 18608228
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608228 100 mL Amber Glass Bottle
18608232 500 mL Amber Glass Bottle
18608231 4 L Amber Glass Bottle
18608229 20 L Steel Can
18608230 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608228 Supplier Spectrum Chemical Mfg Cor Supplier No. M1246100ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 78-78-4

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 mL
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.