missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbiphenyl 95.0+%, TCI America™

Catalog No. p-7132613
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12421G 1 g
M12425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12421G Supplier TCI America Supplier No. M12421G
Only null left

Chemical Identifiers

CAS 643-58-3
Molecular Formula C13H12
Molecular Weight (g/mol) 168.239
MDL Number MFCD00008517
InChI Key ALLIZEAXNXSFGD-UHFFFAOYSA-N
Synonym 2-methylbiphenyl, 2-methyl-1,1'-biphenyl, 2-phenyltoluene, 2-methyl biphenyl, o-phenyltoluene, o-methylbiphenyl, 2-methyl-biphenyl, 1,1'-biphenyl, 2-methyl, biphenyl, 2-methyl, unii-x937mq8s1t
PubChem CID 12563
IUPAC Name 1-methyl-2-phenylbenzene
SMILES CC1=CC=CC=C1C2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 256°C
Quantity 1 g
Formula Weight 168.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylbiphenyl
EINECSNumber (4)-0015
TSCA No